Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAATTTCCACCTCTGTGTGTGTAC[A/G]AGCTCATGGTAAGTGTGACTTTTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 123864 MIM: 604886 | ||||||||||||||||||||
Literature Links: |
COX4I1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COX4I1 - cytochrome c oxidase subunit 4I1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318786.1 | 252 | Missense Mutation | CAA,CGA | Q,R 22 | NP_001305715.1 | |
NM_001318788.1 | 252 | Missense Mutation | CAA,CGA | Q,R 22 | NP_001305717.1 | |
NM_001318794.1 | 252 | Missense Mutation | CAA,CGA | Q,R 22 | NP_001305723.1 | |
NM_001318797.1 | 252 | UTR 5 | NP_001305726.1 | |||
NM_001318802.1 | 252 | UTR 5 | NP_001305731.1 | |||
NM_001861.4 | 252 | Missense Mutation | CAA,CGA | Q,R 22 | NP_001852.1 |
EMC8 - ER membrane protein complex subunit 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC101928557 - uncharacterized LOC101928557 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |