Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCGTGCCGCCCCCTGCAGGGTCT[A/C]CAAGCCCTGGCCCCTCTGAGGGGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114207 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARL5C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARL5C - ADP ribosylation factor like GTPase 5C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CACNB1 - calcium voltage-gated channel auxiliary subunit beta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000723.4 | 2165 | Missense Mutation | GGA,GTA | G,V 533 | NP_000714.3 | |
NM_199247.2 | 2165 | Intron | NP_954855.1 | |||
NM_199248.2 | 2165 | Intron | NP_954856.1 | |||
XM_005257645.2 | 2165 | Missense Mutation | GGA,GTA | G,V 578 | XP_005257702.1 | |
XM_005257646.2 | 2165 | Intron | XP_005257703.1 | |||
XM_006722072.2 | 2165 | Missense Mutation | GGA,GTA | G,V 531 | XP_006722135.1 | |
XM_017025024.1 | 2165 | Missense Mutation | GGA,GTA | G,V 534 | XP_016880513.1 | |
XM_017025025.1 | 2165 | Missense Mutation | GGA,GTA | G,V 486 | XP_016880514.1 | |
XM_017025026.1 | 2165 | Missense Mutation | GGA,GTA | G,V 366 | XP_016880515.1 | |
XM_017025027.1 | 2165 | Missense Mutation | GGA,GTA | G,V 305 | XP_016880516.1 | |
XM_017025028.1 | 2165 | Missense Mutation | GGA,GTA | G,V 305 | XP_016880517.1 | |
XM_017025029.1 | 2165 | Missense Mutation | GGA,GTA | G,V 305 | XP_016880518.1 | |
XM_017025030.1 | 2165 | Intron | XP_016880519.1 |