Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCACGGACAGGTTACTGCAGAGG[A/G]CATCCTCACACATGACCTTTAAGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602650 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPOP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPOP - speckle type BTB/POZ protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007226.1 | 1346 | Missense Mutation | GCC,GTC | A,V 292 | NP_001007227.1 | |
NM_001007227.1 | 1346 | Missense Mutation | GCC,GTC | A,V 292 | NP_001007228.1 | |
NM_001007228.1 | 1346 | Missense Mutation | GCC,GTC | A,V 292 | NP_001007229.1 | |
NM_001007229.1 | 1346 | Missense Mutation | GCC,GTC | A,V 292 | NP_001007230.1 | |
NM_001007230.1 | 1346 | Missense Mutation | GCC,GTC | A,V 292 | NP_001007231.1 | |
NM_003563.3 | 1346 | Missense Mutation | GCC,GTC | A,V 292 | NP_003554.1 | |
XM_005257723.4 | 1346 | Missense Mutation | GCC,GTC | A,V 292 | XP_005257780.1 | |
XM_005257724.4 | 1346 | Missense Mutation | GCC,GTC | A,V 292 | XP_005257781.1 | |
XM_017025204.1 | 1346 | Missense Mutation | GCC,GTC | A,V 292 | XP_016880693.1 |