Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGAAGACAGAGCGGACAGGGCCTT[C/T]GGATCCGCGGGGCTTCCCTGCCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607873 MIM: 610791 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SCARF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SCARF1 - scavenger receptor class F member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003693.3 | 1097 | Missense Mutation | AAA,GAA | K,E 702 | NP_003684.2 | |
NM_145350.2 | 1097 | UTR 3 | NP_663325.1 | |||
XM_017025256.1 | 1097 | Missense Mutation | AAA,GAA | K,E 291 | XP_016880745.1 |
SLC43A2 - solute carrier family 43 member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |