Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGAAGTCCAGCGTGCCCAGGGC[A/G]GCTGCGGACAGAGGAGGGCACAGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604568 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DOC2B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DOC2B - double C2 domain beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003585.4 | 526 | Silent Mutation | ACC,ACT | T,T 125 | NP_003576.2 | |
XM_011535363.2 | 526 | Silent Mutation | ACC,ACT | T,T 125 | XP_011533665.1 | |
XM_017025227.1 | 526 | Silent Mutation | ACC,ACT | T,T 125 | XP_016880716.1 |