Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCCCAGAGGCTGCCCCCCTGCAC[A/G]TTTCCTGTCTCTTACTTGTGACGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605398 MIM: 612383 MIM: 614312 | ||||||||||||||||||||
Literature Links: |
CXCL16 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CXCL16 - C-X-C motif chemokine ligand 16 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MED11 - mediator complex subunit 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZMYND15 - zinc finger MYND-type containing 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136046.2 | 676 | Missense Mutation | ATT,GTT | I,V 204 | NP_001129518.1 | |
NM_001267822.1 | 676 | Missense Mutation | ATT,GTT | I,V 204 | NP_001254751.1 | |
NM_032265.2 | 676 | Missense Mutation | ATT,GTT | I,V 204 | NP_115641.1 | |
XM_017025218.1 | 676 | Intron | XP_016880707.1 |