Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTGACCACCCTGGCCTCAACTGA[G/T]GTTGGGGTTTGATCCTGGGGGAGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601333 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PROCA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PROCA1 - protein interacting with cyclin A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304949.1 | 1217 | Missense Mutation | ATC,CTC | I,L 326 | NP_001291878.1 | |
NM_001304951.1 | 1217 | Missense Mutation | ATC,CTC | I,L 219 | NP_001291880.1 | |
NM_001304952.1 | 1217 | Missense Mutation | ATC,CTC | I,L 337 | NP_001291881.1 | |
NM_001304953.1 | 1217 | Missense Mutation | ATC,CTC | I,L 263 | NP_001291882.1 | |
NM_001304954.1 | 1217 | Missense Mutation | ATC,CTC | I,L 263 | NP_001291883.1 | |
NM_152465.2 | 1217 | Missense Mutation | ATC,CTC | I,L 335 | NP_689678.1 | |
XM_005257927.4 | 1217 | Missense Mutation | ATC,CTC | I,L 363 | XP_005257984.1 | |
XM_006721723.3 | 1217 | Missense Mutation | ATC,CTC | I,L 360 | XP_006721786.1 | |
XM_011524398.2 | 1217 | Missense Mutation | ATC,CTC | I,L 354 | XP_011522700.1 | |
XM_011524399.2 | 1217 | Missense Mutation | ATC,CTC | I,L 326 | XP_011522701.1 | |
XM_011524400.2 | 1217 | Missense Mutation | ATC,CTC | I,L 309 | XP_011522702.1 | |
XM_011524401.1 | 1217 | Missense Mutation | ATC,CTC | I,L 263 | XP_011522703.1 | |
XM_017024237.1 | 1217 | Missense Mutation | ATC,CTC | I,L 263 | XP_016879726.1 |
SUPT6H - SPT6 homolog, histone chaperone | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320755.1 | 1217 | Intron | NP_001307684.1 | |||
NM_003170.4 | 1217 | Intron | NP_003161.2 | |||
XM_017024978.1 | 1217 | Intron | XP_016880467.1 | |||
XM_017024979.1 | 1217 | Intron | XP_016880468.1 | |||
XM_017024980.1 | 1217 | Intron | XP_016880469.1 |