Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCCTCTTCCTCTTCTTCCTCTTC[C/T]TCTTCTTCCTCTTCTTCCTCTTCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609455 | ||||||||||||||||||||
Literature Links: |
PELP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PELP1 - proline, glutamate and leucine rich protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278241.1 | 2923 | Silent Mutation | GAA,GAG | E,E 752 | NP_001265170.1 | |
NM_014389.2 | 2923 | Silent Mutation | GAA,GAG | E,E 949 | NP_055204.3 |