Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCGGCGGAGAAGCCTGATGAGGC[C/G]TATCTCCTGAAGTTGGAAAGCAATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611825 MIM: 606735 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRC46 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRRC46 - leucine rich repeat containing 46 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPL10 - mitochondrial ribosomal protein L10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145255.3 | 357 | Missense Mutation | CGC,GGC | R,G 77 | NP_660298.2 | |
NM_148887.2 | 357 | Missense Mutation | CGC,GGC | R,G 87 | NP_683685.1 |
OSBPL7 - oxysterol binding protein like 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |