Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607435 MIM: 612070 MIM: 612071 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERAL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ERAL1 - Era like 12S mitochondrial rRNA chaperone 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317985.1 | 82 | Missense Mutation | CCT,GCT | P,A 2 | NP_001304914.1 | |
NM_001317986.1 | 82 | Missense Mutation | CCT,GCT | P,A 2 | NP_001304915.1 | |
NM_005702.3 | 82 | Missense Mutation | CCT,GCT | P,A 2 | NP_005693.1 |
FAM222B - family with sequence similarity 222 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077498.2 | 82 | Intron | NP_001070966.1 | |||
NM_001288631.1 | 82 | Intron | NP_001275560.1 | |||
NM_001288632.1 | 82 | Intron | NP_001275561.1 | |||
NM_001288633.1 | 82 | Intron | NP_001275562.1 | |||
NM_001288634.1 | 82 | Intron | NP_001275563.1 | |||
NM_001288635.1 | 82 | Intron | NP_001275564.1 | |||
NM_001288636.1 | 82 | Intron | NP_001275565.1 | |||
NM_001288637.1 | 82 | Intron | NP_001275566.1 | |||
NM_001288638.1 | 82 | Intron | NP_001275567.1 | |||
NM_001288639.1 | 82 | Intron | NP_001275568.1 | |||
NM_001288640.1 | 82 | Intron | NP_001275569.1 | |||
NM_018182.3 | 82 | Intron | NP_060652.2 | |||
XM_011524986.2 | 82 | Intron | XP_011523288.1 | |||
XM_011524987.1 | 82 | Intron | XP_011523289.1 | |||
XM_011524988.2 | 82 | Intron | XP_011523290.1 | |||
XM_017024831.1 | 82 | Intron | XP_016880320.1 | |||
XM_017024832.1 | 82 | Intron | XP_016880321.1 | |||
XM_017024833.1 | 82 | Intron | XP_016880322.1 | |||
XM_017024834.1 | 82 | Intron | XP_016880323.1 | |||
XM_017024835.1 | 82 | Intron | XP_016880324.1 |
MIR144 - microRNA 144 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR451A - microRNA 451a | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR451B - microRNA 451b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4732 - microRNA 4732 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |