Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCAAAATGATGCTTTTTCAGTAT[C/T]TGCAGGGATTTCAACATCGTCATAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614406 | ||||||||||||||||||||
Literature Links: |
LOC100130950 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC100130950 - uncharacterized LOC100130950 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCIMP - SLP adaptor and CSK interacting membrane protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271842.1 | 520 | Missense Mutation | AAT,GAT | N,D 132 | NP_001258771.1 | |
NM_001319190.1 | 520 | Intron | NP_001306119.1 | |||
NM_207103.3 | 520 | Missense Mutation | AAT,GAT | N,D 139 | NP_996986.1 | |
XM_017024615.1 | 520 | Intron | XP_016880104.1 |