Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGTCCTACTCTCCGCGGTGCTCTG[C/G]TGCCTCACAGGAGGTGGGGCTCCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606221 MIM: 608499 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IKZF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IKZF3 - IKAROS family zinc finger 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZPBP2 - zona pellucida binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198844.2 | 192 | Missense Mutation | TGC,TGG | C,W 13 | NP_942141.2 | |
NM_199321.2 | 192 | Missense Mutation | TGC,TGG | C,W 13 | NP_955353.1 | |
XM_011524298.1 | 192 | Missense Mutation | TGC,TGG | C,W 13 | XP_011522600.1 |