Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCAAACCTTCCATTGAACCCATC[A/G]ATCGGTTTCAGATCAGCATTGTAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C17orf98 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C17orf98 - chromosome 17 open reading frame 98 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080465.2 | 366 | Silent Mutation | ATC,ATT | I,I 122 | NP_001073934.1 |
CWC25 - CWC25 spliceosome associated protein homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4727 - microRNA 4727 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |