Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGAATTTGGGGTTTTAAGGCTCT[G/T]CCTGGGGGAGGAGCTTCCACTTCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612505 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCA8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABCA8 - ATP binding cassette subfamily A member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288985.1 | 4968 | Missense Mutation | NP_001275914.1 | |||
NM_001288986.1 | 4968 | Missense Mutation | NP_001275915.1 | |||
NM_007168.3 | 4968 | Missense Mutation | NP_009099.1 | |||
XM_005256938.2 | 4968 | Missense Mutation | XP_005256995.1 | |||
XM_005256940.4 | 4968 | Missense Mutation | XP_005256997.1 | |||
XM_011524191.2 | 4968 | Intron | XP_011522493.1 | |||
XM_011524192.2 | 4968 | Intron | XP_011522494.1 |