Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGCGCCGAGGCGGCCGAGGGCCA[G/T]TTCGACCGCTACTACCACGAAGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603102 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CPD - carboxypeptidase D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199775.1 | 219 | Intron | NP_001186704.1 | |||
NM_001304.4 | 219 | Missense Mutation | CAG,CAT | Q,H 54 | NP_001295.2 |