Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTATAGACCAACAGCAAAGCCTTTA[C/T]TGCCAAGACTTCCTGTGTGCGGCGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614906 | ||||||||||||||||||||
Literature Links: |
SNX11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SNX11 - sorting nexin 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013323.2 | 416 | Missense Mutation | ACT,ATT | T,I 50 | NP_037455.2 | |
NM_152244.1 | 416 | Missense Mutation | ACT,ATT | T,I 50 | NP_689450.1 | |
XM_005257260.3 | 416 | Missense Mutation | ACT,ATT | T,I 50 | XP_005257317.1 | |
XM_005257261.3 | 416 | Missense Mutation | ACT,ATT | T,I 42 | XP_005257318.1 | |
XM_005257262.3 | 416 | Missense Mutation | ACT,ATT | T,I 42 | XP_005257319.1 | |
XM_011524697.2 | 416 | Missense Mutation | ACT,ATT | T,I 50 | XP_011522999.1 |