Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCCGGGGCCCGAGAACAAGTACGA[C/T]CCGGAGAAGGACAAGACCAACTTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
SHISA6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SHISA6 - shisa family member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001173461.1 | 504 | Silent Mutation | GAC,GAT | D,D 168 | NP_001166932.1 | |
NM_001173462.1 | 504 | Silent Mutation | GAC,GAT | D,D 168 | NP_001166933.1 | |
NM_207386.3 | 504 | Silent Mutation | GAC,GAT | D,D 168 | NP_997269.2 | |
XM_017024618.1 | 504 | Silent Mutation | GAC,GAT | D,D 168 | XP_016880107.1 |