Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTCTGCTGAGGCTCCCCATCCTC[C/T]AGGCCGCCTGCCAGCTCCCTGGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605367 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP44 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGAP44 - Rho GTPase activating protein 44 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ELAC2 - elaC ribonuclease Z 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001165962.1 | 2523 | Silent Mutation | CTA,CTG | L,L 763 | NP_001159434.1 | |
NM_018127.6 | 2523 | Silent Mutation | CTA,CTG | L,L 803 | NP_060597.4 | |
NM_173717.1 | 2523 | Silent Mutation | CTA,CTG | L,L 802 | NP_776065.1 |