Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGGCAGTGGTCAAGTGTCTGTTC[A/G]AGAACGTCCAGTGAGTATGAGCGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138033 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GCGR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GCGR - glucagon receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000160.4 | 1163 | Missense Mutation | AAG,GAG | K,E 290 | NP_000151.1 | |
XM_006722277.1 | 1163 | Missense Mutation | AAG,GAG | K,E 290 | XP_006722340.1 | |
XM_011523539.1 | 1163 | Missense Mutation | AAG,GAG | K,E 257 | XP_011521841.1 | |
XM_017024446.1 | 1163 | Missense Mutation | AAG,GAG | K,E 288 | XP_016879935.1 | |
XM_017024447.1 | 1163 | Missense Mutation | AAG,GAG | K,E 168 | XP_016879936.1 |