Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTCTCTGCTAAAGACTGCGTTCA[C/T]GTCCACCTGGATCTGACCTGGTGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607827 MIM: 607828 MIM: 607696 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OTOP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OTOP2 - otopetrin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178160.2 | 581 | Silent Mutation | CAC,CAT | H,H 163 | NP_835454.1 | |
XM_011525479.1 | 581 | Silent Mutation | CAC,CAT | H,H 163 | XP_011523781.1 |
OTOP3 - otopetrin 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USH1G - USH1 protein network component sans | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |