Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGTTTCCCGCAGTCGCGGGCAGGG[A/C]GCCAAGGCGGCAGGAGGAGGGTGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607570 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DHX40 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DHX40 - DEAH-box helicase 40 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166301.1 | 125 | Missense Mutation | GAG,GCG | E,A 11 | NP_001159773.1 | |
NM_024612.4 | 125 | Missense Mutation | GAG,GCG | E,A 11 | NP_078888.4 | |
XM_006722083.2 | 125 | Missense Mutation | GAG,GCG | E,A 11 | XP_006722146.1 | |
XM_011525253.2 | 125 | Missense Mutation | GAG,GCG | E,A 11 | XP_011523555.1 | |
XM_011525254.2 | 125 | UTR 5 | XP_011523556.1 | |||
XM_017025080.1 | 125 | Missense Mutation | GAG,GCG | E,A 11 | XP_016880569.1 | |
XM_017025081.1 | 125 | UTR 5 | XP_016880570.1 | |||
XM_017025082.1 | 125 | UTR 5 | XP_016880571.1 | |||
XM_017025083.1 | 125 | UTR 5 | XP_016880572.1 |