Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGCAGCTGCTCCTTGGTGAAGGCC[A/G]TCCTCTCCTTGCGGGCTTTGCTGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600147 | ||||||||||||||||||||
Literature Links: |
MEOX1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MEOX1 - mesenchyme homeobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040002.1 | 947 | Missense Mutation | ACG,ATG | T,M 61 | NP_001035091.1 | |
NM_004527.3 | 947 | Missense Mutation | ACG,ATG | T,M 176 | NP_004518.1 | |
NM_013999.3 | 947 | Intron | NP_054705.1 | |||
XM_011524818.1 | 947 | Missense Mutation | ACG,ATG | T,M 176 | XP_011523120.1 |