Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCTGAGATGGGGGGCAGGTTGCA[C/T]CTGCAGAAGAGAAAAGGCAGCCCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607162 | ||||||||||||||||||||
Literature Links: |
ST8SIA5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ST8SIA5 - ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001307986.1 | 1339 | Silent Mutation | NP_001294915.1 | |||
NM_001307987.1 | 1339 | Silent Mutation | NP_001294916.1 | |||
NM_013305.5 | 1339 | Silent Mutation | NP_037437.2 | |||
XM_017025715.1 | 1339 | Silent Mutation | XP_016881204.1 | |||
XM_017025716.1 | 1339 | Silent Mutation | XP_016881205.1 |