Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCTTCATAGTTATACGAACAAAC[A/G]TAGTCTTCACAATGTTTATGCTCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 125643 | ||||||||||||||||||||
Literature Links: |
DSC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DSC1 - desmocollin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004948.3 | 2800 | UTR 3 | NP_004939.1 | |||
NM_024421.2 | 2800 | Silent Mutation | TAC,TAT | Y,Y 846 | NP_077739.1 |
DSCAS - DSC1/DSC2 antisense RNA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |