Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTGACATGGATCTTCTCCAGGAC[A/G]AATGCTGTTGTTCACAATACTATCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF563 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF563 - zinc finger protein 563 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145276.2 | 527 | Missense Mutation | CGT,TGT | R,C 97 | NP_660319.1 | |
XM_005259750.4 | 527 | Intron | XP_005259807.1 | |||
XM_005259751.3 | 527 | Intron | XP_005259808.1 | |||
XM_006722650.3 | 527 | Intron | XP_006722713.1 | |||
XM_006722651.3 | 527 | Intron | XP_006722714.1 | |||
XM_011527698.1 | 527 | Intron | XP_011526000.1 | |||
XM_011527699.2 | 527 | Missense Mutation | CGT,TGT | R,C 60 | XP_011526001.1 | |
XM_011527700.2 | 527 | Intron | XP_011526002.1 | |||
XM_017026332.1 | 527 | Intron | XP_016881821.1 |