Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACTCAGCGTGTCATACCTGGTGG[C/T]CAGCTACATCGTGAGCACAGGCGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614591 | ||||||||||||||||||||
Literature Links: |
CEACAM16 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CEACAM16 - carcinoembryonic antigen related cell adhesion molecule 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039213.3 | 403 | Missense Mutation | GCC,GTC | A,V 66 | NP_001034302.2 | |
XM_017026795.1 | 403 | Missense Mutation | GCC,GTC | A,V 66 | XP_016882284.1 |