Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCGATGGGGCTGGATATAGTTCC[C/T]CAGCCTGACACTCGGCAGGAGGTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606135 | ||||||||||||||||||||
Literature Links: |
KLK14 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KLK14 - kallikrein related peptidase 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001311182.1 | 676 | Nonsense Mutation | TGA,TGG | *,W 164 | NP_001298111.1 | |
NM_022046.5 | 676 | Nonsense Mutation | TGA,TGG | *,W 164 | NP_071329.2 | |
XM_017026820.1 | 676 | Nonsense Mutation | TGA,TGG | *,W 164 | XP_016882309.1 |