Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAACAGCTGCCACAGCACGCTCTC[A/G]TCTGCCTCCGGTGACAGGTTGTACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603458 MIM: 177060 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ELAVL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ELAVL3 - ELAV like neuron-specific RNA binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001420.3 | 1271 | Silent Mutation | GAC,GAT | D,D 296 | NP_001411.2 | |
NM_032281.2 | 1271 | Silent Mutation | GAC,GAT | D,D 289 | NP_115657.2 | |
XM_011527778.2 | 1271 | Silent Mutation | GAC,GAT | D,D 295 | XP_011526080.1 |
PRKCSH - protein kinase C substrate 80K-H | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |