Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGTCTGAATAATTTCCCACATTC[A/G]CCACACTCATATGGCTTTTCTCCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF416 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF416 - zinc finger protein 416 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017879.1 | 1326 | Silent Mutation | GGC,GGT | G,G 385 | NP_060349.1 | |
XM_005259051.3 | 1326 | Silent Mutation | GGC,GGT | G,G 313 | XP_005259108.1 |