Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAGGGGGTCCTGGGCCCACCTGG[A/T]GCATGATCTTGGTGAAGGCCTCCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614194 MIM: 614610 | ||||||||||||||||||||
Literature Links: |
DOCK6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DOCK6 - dedicator of cytokinesis 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020812.3 | 5310 | Missense Mutation | CAC,CTC | H,L 1739 | NP_065863.2 | |
XM_005260000.2 | 5310 | Missense Mutation | CAC,CTC | H,L 1805 | XP_005260057.1 | |
XM_005260001.2 | 5310 | Missense Mutation | CAC,CTC | H,L 1774 | XP_005260058.1 | |
XM_006722804.3 | 5310 | Missense Mutation | CAC,CTC | H,L 851 | XP_006722867.1 | |
XM_011528150.1 | 5310 | Missense Mutation | CAC,CTC | H,L 1785 | XP_011526452.1 | |
XM_011528151.1 | 5310 | Missense Mutation | CAC,CTC | H,L 1781 | XP_011526453.1 | |
XM_011528152.1 | 5310 | Missense Mutation | CAC,CTC | H,L 1750 | XP_011526454.1 |
KANK2 - KN motif and ankyrin repeat domains 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105372273 - uncharacterized LOC105372273 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |