Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGGCTGAAGGCTCTTCCACACTG[A/G]TTACATTCAAATGGTTTTTCTCCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613749 | ||||||||||||||||||||
Literature Links: |
ZNF260 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF260 - zinc finger protein 260 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012756.2 | 1417 | Silent Mutation | AAC,AAT | N,N 167 | NP_001012774.1 | |
NM_001166036.1 | 1417 | Silent Mutation | AAC,AAT | N,N 167 | NP_001159508.1 | |
NM_001166037.1 | 1417 | Silent Mutation | AAC,AAT | N,N 167 | NP_001159509.1 | |
NM_001166038.1 | 1417 | Silent Mutation | AAC,AAT | N,N 167 | NP_001159510.1 | |
XM_011526897.2 | 1417 | Silent Mutation | AAC,AAT | N,N 167 | XP_011525199.1 | |
XM_017026741.1 | 1417 | Silent Mutation | AAC,AAT | N,N 167 | XP_016882230.1 |