Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCAATGCCCAGGCCCAGGCCGAT[A/G]CCCAGGGGGATCATGGCGCTTACGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612168 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC39A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC39A3 - solute carrier family 39 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144564.4 | 983 | Silent Mutation | GGC,GGT | G,G 243 | NP_653165.2 | |
NM_213568.1 | 983 | Intron | NP_998733.1 |