Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACGGGTCCCCAGGCCTTCAGCCGC[A/G]TCCAGATCTACCACAACCCCACGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601703 | ||||||||||||||||||||
Literature Links: |
VASP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
VASP - vasodilator-stimulated phosphoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003370.3 | 436 | Missense Mutation | ATC,GTC | I,V 36 | NP_003361.1 | |
XM_005259199.1 | 436 | Missense Mutation | ATC,GTC | I,V 36 | XP_005259256.1 | |
XM_005259200.1 | 436 | Missense Mutation | ATC,GTC | I,V 36 | XP_005259257.1 | |
XM_017027200.1 | 436 | Missense Mutation | ATC,GTC | I,V 36 | XP_016882689.1 |