Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCCTTACACTCAAAGGGTTTTTC[A/G]CCAGTATGAATGCTCTGATGTAGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZFP82 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZFP82 - ZFP82 zinc finger protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321917.1 | 1683 | Silent Mutation | GGC,GGT | G,G 475 | NP_001308846.1 | |
NM_001321918.1 | 1683 | Intron | NP_001308847.1 | |||
NM_001321919.1 | 1683 | Intron | NP_001308848.1 | |||
NM_133466.3 | 1683 | Silent Mutation | GGC,GGT | G,G 474 | NP_597723.1 |