Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATGGCCTGGAAGTAGATGACACC[A/T]TTGAGCTTGGTCTCTTCCTTGTCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
PHLDB3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PHLDB3 - pleckstrin homology like domain family B member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198850.3 | 2156 | Missense Mutation | AAA,AAT | K,N 575 | NP_942147.3 | |
XM_005259169.3 | 2156 | Missense Mutation | AAA,AAT | K,N 570 | XP_005259226.1 | |
XM_005259170.3 | 2156 | Missense Mutation | AAA,AAT | K,N 536 | XP_005259227.1 | |
XM_005259171.3 | 2156 | Intron | XP_005259228.1 | |||
XM_011527236.2 | 2156 | Missense Mutation | AAA,AAT | K,N 570 | XP_011525538.1 |