Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACACAGATCACCCTGGGAATCCT[C/T]ATCTCCACTCAGCGGGCCCAGGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 151523 MIM: 607972 MIM: 601729 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD37 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD37 - CD37 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040031.1 | 440 | Silent Mutation | CTC,CTT | L,L 39 | NP_001035120.1 | |
NM_001774.2 | 440 | Silent Mutation | CTC,CTT | L,L 107 | NP_001765.1 | |
XM_005259435.3 | 440 | Silent Mutation | CTC,CTT | L,L 107 | XP_005259492.1 | |
XM_005259436.3 | 440 | Silent Mutation | CTC,CTT | L,L 39 | XP_005259493.1 | |
XM_011527542.2 | 440 | Silent Mutation | CTC,CTT | L,L 39 | XP_011525844.1 | |
XM_011527543.2 | 440 | Silent Mutation | CTC,CTT | L,L 39 | XP_011525845.1 | |
XM_011527544.2 | 440 | Silent Mutation | CTC,CTT | L,L 39 | XP_011525846.1 | |
XM_017027513.1 | 440 | Silent Mutation | CTC,CTT | L,L 39 | XP_016883002.1 |
SLC6A16 - solute carrier family 6 member 16 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEAD2 - TEA domain transcription factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |