Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGCCTGGCTTGCCACGTGATGGGG[A/G]CTCTGCCCTCCCTAGCTTGGGCATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611712 MIM: 605725 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HIPK4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HIPK4 - homeodomain interacting protein kinase 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRX - periaxin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020956.2 | 3358 | UTR 3 | NP_066007.1 | |||
NM_181882.2 | 3358 | Missense Mutation | CCC,TCC | P,S 814 | NP_870998.2 | |
XM_011527171.2 | 3358 | Missense Mutation | CCC,TCC | P,S 814 | XP_011525473.1 | |
XM_017027046.1 | 3358 | Missense Mutation | CCC,TCC | P,S 780 | XP_016882535.1 | |
XM_017027047.1 | 3358 | Missense Mutation | CCC,TCC | P,S 780 | XP_016882536.1 |