Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTCTCCTGGGGGCCTGGTGGTCC[C/T]AGGCAGGGGGCCGGGCTGGGGACTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605343 | ||||||||||||||||||||
Literature Links: |
FSTL3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FSTL3 - follistatin like 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRSS57 - protease, serine 57 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308209.1 | 848 | Missense Mutation | AGG,GGG | R,G 273 | NP_001295138.1 | |
NM_214710.4 | 848 | Missense Mutation | AGG,GGG | R,G 274 | NP_999875.1 |