Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTGGGCCTTGGTCACTTCTGCTT[C/G]TAGCCCAGGCTCGGTCACCATGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609171 MIM: 616575 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDC42EP5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDC42EP5 - CDC42 effector protein 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LENG8 - leukocyte receptor cluster member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052925.3 | 1119 | Intron | NP_443157.1 | |||
XM_005278248.4 | 1119 | Intron | XP_005278305.1 | |||
XM_005278249.4 | 1119 | Intron | XP_005278306.1 | |||
XM_005278250.4 | 1119 | Intron | XP_005278307.1 | |||
XM_005278252.4 | 1119 | Intron | XP_005278309.1 | |||
XM_006722996.3 | 1119 | Intron | XP_006723059.1 | |||
XM_006722997.3 | 1119 | Intron | XP_006723060.1 | |||
XM_011526413.2 | 1119 | Intron | XP_011524715.1 | |||
XM_011526414.2 | 1119 | Intron | XP_011524716.1 | |||
XM_011526415.2 | 1119 | Intron | XP_011524717.1 |
LENG9 - leukocyte receptor cluster member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301782.1 | 1119 | Missense Mutation | CAA,GAA | Q,E 311 | NP_001288711.1 | |
NM_198988.1 | 1119 | Missense Mutation | CAA,GAA | Q,E 334 | NP_945339.2 |