Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGTACCTGGCAGAATGTTTCCTT[A/G]TGGACTTTGCCAAGAACATTCCAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TDRD12 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TDRD12 - tudor domain containing 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001110822.1 | 343 | Missense Mutation | ATG,GTG | M,V 95 | NP_001104292.1 | |
XM_011527471.2 | 343 | Missense Mutation | ATG,GTG | M,V 95 | XP_011525773.1 | |
XM_011527472.2 | 343 | Missense Mutation | ATG,GTG | M,V 95 | XP_011525774.1 | |
XM_011527473.2 | 343 | Missense Mutation | ATG,GTG | M,V 95 | XP_011525775.1 | |
XM_011527474.2 | 343 | Missense Mutation | ATG,GTG | M,V 95 | XP_011525776.1 | |
XM_011527475.2 | 343 | Missense Mutation | ATG,GTG | M,V 95 | XP_011525777.1 | |
XM_011527476.2 | 343 | Missense Mutation | ATG,GTG | M,V 95 | XP_011525778.1 | |
XM_017027458.1 | 343 | Missense Mutation | ATG,GTG | M,V 95 | XP_016882947.1 | |
XM_017027459.1 | 343 | Intron | XP_016882948.1 |