Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCACATTCGTTACATTCATAAGG[C/T]TTCTCACCCGTGTGGGTCCTCTGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 611703 | ||||||||||||||||||||
Literature Links: |
ZNF436 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF436 - zinc finger protein 436 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077195.1 | 1387 | Silent Mutation | AAA,AAG | K,K 276 | NP_001070663.1 | |
NM_030634.2 | 1387 | Silent Mutation | AAA,AAG | K,K 276 | NP_085137.1 | |
XM_011542201.1 | 1387 | Silent Mutation | AAA,AAG | K,K 258 | XP_011540503.1 |
ZNF436-AS1 - ZNF436 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |