Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGACTCTCCATGGATGGGGAGGA[G/T]GCCCCAGCCCCTGAGGACGTCACCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610770 MIM: 616765 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOC2L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NOC2L - NOC2 like nucleolar associated transcriptional repressor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015658.3 | 1679 | Intron | NP_056473.2 |
SAMD11 - sterile alpha motif domain containing 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152486.2 | 1679 | Missense Mutation | GAG,GAT | E,D 533 | NP_689699.2 |