Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGGTGGCAGGGGTCAGGGGCTGG[C/G]TTCCTCAGGCAGCTGGCCTGCATAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616597 MIM: 176889 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARL8A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARL8A - ADP ribosylation factor like GTPase 8A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTPN7 - protein tyrosine phosphatase, non-receptor type 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199797.1 | 1345 | Missense Mutation | CCC,GCC | P,A 432 | NP_001186726.1 | |
NM_002832.3 | 1345 | Missense Mutation | CCC,GCC | P,A 463 | NP_002823.3 | |
NM_080588.2 | 1345 | Missense Mutation | CCC,GCC | P,A 397 | NP_542155.1 | |
XM_011509819.1 | 1345 | Missense Mutation | CCC,GCC | P,A 509 | XP_011508121.1 | |
XM_011509820.2 | 1345 | Missense Mutation | CCC,GCC | P,A 398 | XP_011508122.1 | |
XM_011509821.1 | 1345 | Missense Mutation | CCC,GCC | P,A 352 | XP_011508123.1 | |
XM_011509822.2 | 1345 | Missense Mutation | CCC,GCC | P,A 352 | XP_011508124.1 | |
XM_011509823.1 | 1345 | Intron | XP_011508125.1 | |||
XM_011509824.1 | 1345 | Missense Mutation | CCC,GCC | P,A 233 | XP_011508126.1 | |
XM_017001935.1 | 1345 | Missense Mutation | CCC,GCC | P,A 508 | XP_016857424.1 | |
XM_017001936.1 | 1345 | Missense Mutation | CCC,GCC | P,A 462 | XP_016857425.1 | |
XM_017001937.1 | 1345 | Missense Mutation | CCC,GCC | P,A 357 | XP_016857426.1 |