Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAGCAGATCAATATCACCCTGGA[C/T]CACCGCTGCCGTATCTTCCAGAACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 153454 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLOD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLOD1 - procollagen-lysine,2-oxoglutarate 5-dioxygenase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000302.3 | 857 | Silent Mutation | GAC,GAT | D,D 201 | NP_000293.2 | |
NM_001316320.1 | 857 | Silent Mutation | GAC,GAT | D,D 248 | NP_001303249.1 |