Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCCAAGGCATCTCCAAGGACATG[A/C]AGAAAAACTCCACGCATGTTAAGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 609521 | ||||||||||||||||||||
Literature Links: |
SLC30A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC30A1 - solute carrier family 30 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021194.2 | 895 | Silent Mutation | CTG,CTT | L,L 250 | NP_067017.2 |