Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGCCCACAGCCCGATTTCATCAG[A/C]CATTAGGTCGATGTCAGAGAATCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607567 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OLFM3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OLFM3 - olfactomedin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288821.1 | 1313 | Missense Mutation | GCT,TCT | A,S 351 | NP_001275750.1 | |
NM_001288823.1 | 1313 | Missense Mutation | GCT,TCT | A,S 256 | NP_001275752.1 | |
NM_058170.3 | 1313 | Missense Mutation | GCT,TCT | A,S 331 | NP_477518.2 | |
XM_017000240.1 | 1313 | Missense Mutation | GCT,TCT | A,S 256 | XP_016855729.1 |