Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCTGGGCTCTGATGTTGAACTTA[G/T]ATGCCGGTGCCTCTTCTTCCTCCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 616554 MIM: 604737 | ||||||||||||||||||||
Literature Links: |
SPAG17 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SPAG17 - sperm associated antigen 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_206996.2 | 2621 | Intron | NP_996879.1 | |||
XM_006710426.3 | 2621 | Intron | XP_006710489.1 | |||
XM_006710427.3 | 2621 | Intron | XP_006710490.1 | |||
XM_006710428.3 | 2621 | Intron | XP_006710491.1 | |||
XM_006710429.3 | 2621 | Intron | XP_006710492.1 | |||
XM_006710431.3 | 2621 | Intron | XP_006710494.1 | |||
XM_011540934.1 | 2621 | Intron | XP_011539236.1 | |||
XM_011540935.2 | 2621 | Intron | XP_011539237.1 | |||
XM_011540936.2 | 2621 | Intron | XP_011539238.1 | |||
XM_011540937.2 | 2621 | Intron | XP_011539239.1 | |||
XM_011540939.2 | 2621 | Intron | XP_011539241.1 | |||
XM_011540941.2 | 2621 | Intron | XP_011539243.1 | |||
XM_011540942.2 | 2621 | Intron | XP_011539244.1 |
WDR3 - WD repeat domain 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006784.2 | 2621 | Missense Mutation | AGA,ATA | R,I 852 | NP_006775.1 |