Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTGGCTCCTTGCAGCAGGAGTCC[A/G]CATCAGGGGCTGGGGAGGTGGCCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616758 MIM: 610325 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KDF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KDF1 - keratinocyte differentiation factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152365.2 | 644 | Missense Mutation | NP_689578.2 | |||
XM_005245735.2 | 644 | Missense Mutation | XP_005245792.1 | |||
XM_011540622.1 | 644 | Missense Mutation | XP_011538924.1 |
NUDC - nuclear distribution C, dynein complex regulator | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |