Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCACAACCCTTAATACCATCTTCA[A/T]AAATGTGAACAACAATTGTGGTGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 171490 | ||||||||||||||||||||
Literature Links: |
LOC102724919 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC102724919 - uncharacterized LOC102724919 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDC - phosducin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002597.4 | 496 | Missense Mutation | TAT,TTT | Y,F 141 | NP_002588.3 | |
NM_022576.3 | 496 | Missense Mutation | TAT,TTT | Y,F 89 | NP_072098.1 | |
XM_011509603.2 | 496 | Missense Mutation | TAT,TTT | Y,F 141 | XP_011507905.1 |