Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTAAATACGGTCAACAACTTTGG[A/C]CCTACTTCGTACATCCTACAAAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605481 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASPM PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ASPM - abnormal spindle microtubule assembly | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206846.1 | 10434 | Missense Mutation | GCC,TCC | A,S 1808 | NP_001193775.1 | |
NM_018136.4 | 10434 | Missense Mutation | GCC,TCC | A,S 3393 | NP_060606.3 |